Product Details

SNP ID
rs146075107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5106129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTCTGATGCTGACCGAGGTTTG[A/C]GCTCTGTTTGAAAGTTTTCCCACAG
Phenotype
MIM: 194480 MIM: 616463
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928000 PubMed Links

Gene Details

Gene
LOC101928000
Gene Name
uncharacterized LOC101928000
There are no transcripts associated with this gene.

Gene
ZFP3
Gene Name
ZFP3 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZNF232
Gene Name
zinc finger protein 232
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320952.1 1148 Missense Mutation GCA,TCA A335S NP_001307881.1
NM_001320953.1 1148 Missense Mutation GCA,TCA A317S NP_001307882.1
NM_001320954.1 1148 Missense Mutation GCA,TCA A317S NP_001307883.1
NM_001320955.1 1148 Missense Mutation GCA,TCA A316S NP_001307884.1
NM_014519.3 1148 Missense Mutation GCA,TCA A344S NP_055334.2
XM_011524006.2 1148 Missense Mutation GCA,TCA A296S XP_011522308.1
XM_017025021.1 1148 Missense Mutation GCA,TCA A344S XP_016880510.1
XM_017025022.1 1148 Missense Mutation GCA,TCA A268S XP_016880511.1
XM_017025023.1 1148 Missense Mutation GCA,TCA A259S XP_016880512.1

View Full Product Details