Product Details

SNP ID
rs146238052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78111159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTAGGTGTTCACCACATTCCAG[C/G]AGCACACGATCACAGCAACACCTCG
Phenotype
MIM: 605828 MIM: 610741
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMC6 PubMed Links

Gene Details

Gene
TMC6
Gene Name
transmembrane channel like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127198.2 Intron NP_001120670.1
NM_001321185.1 Intron NP_001308114.1
NM_007267.7 Intron NP_009198.4
XM_005256995.1 Intron XP_005257052.1
XM_011524255.1 Intron XP_011522557.1
XM_011524256.1 Intron XP_011522558.1
XM_011524257.2 Intron XP_011522559.1
XM_011524258.1 Intron XP_011522560.1
XM_017024107.1 Intron XP_016879596.1
XM_017024108.1 Intron XP_016879597.1
XM_017024109.1 Intron XP_016879598.1
Gene
TNRC6C
Gene Name
trinucleotide repeat containing 6C
There are no transcripts associated with this gene.

Gene
TNRC6C-AS1
Gene Name
TNRC6C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details