Product Details

SNP ID
rs146522350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49002901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAACTGTTCTCATTTGGGTGGTCA[G/A]TATTTCAAGATGAGAGTTTTGTAAG
Phenotype
MIM: 608288
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
IGF2BP1 PubMed Links

Gene Details

Gene
IGF2BP1
Gene Name
insulin like growth factor 2 mRNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160423.1 Intron NP_001153895.1
NM_006546.3 Intron NP_006537.3
XM_005256955.3 Intron XP_005257012.2
XM_011524201.2 Intron XP_011522503.1
XM_017024022.1 Intron XP_016879511.1

View Full Product Details