Product Details

SNP ID
rs147323689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36088733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCACTCCTCACTGGGGTCAGCA[C/G]AGACCTGCCGGCTTCGCTTGGTTAG
Phenotype
MIM: 182283
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCL3 PubMed Links

Gene Details

Gene
CCL3
Gene Name
C-C motif chemokine ligand 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002983.2 320 Missense Mutation TCT,TGT S73C NP_002974.1

View Full Product Details