Product Details

SNP ID
rs147655478
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27878392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATATATTTACTTCATTGCCAGC[G/A]GAGTACACTCGCATACATAGATATC
Phenotype
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
LYRM9 PubMed Links

Gene Details

Gene
LYRM9
Gene Name
LYR motif containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076680.1 1403 UTR 3 NP_001070148.1
XM_011524491.2 1403 UTR 3 XP_011522793.1
XM_017024325.1 1403 UTR 3 XP_016879814.1

View Full Product Details