Product Details

SNP ID
rs148696353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78971688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGTGCTCTTCGAGCTGTTGGTG[G/T]CCAGGGCACTGGGAATCGCAGCCTT
Phenotype
MIM: 600626
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LGALS3BP PubMed Links

Gene Details

Gene
LGALS3BP
Gene Name
galectin 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005567.3 1825 Missense Mutation GAC,GCC D549A NP_005558.1

View Full Product Details