Product Details

SNP ID
rs150313949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41817065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCCTCCCCCCCCAGCTATGAT[C/T]GCAACACCTTGGTGGCCATCGTGGT
Phenotype
MIM: 607063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP10 PubMed Links

Gene Details

Gene
FKBP10
Gene Name
FK506 binding protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021939.3 578 Missense Mutation CGC,TGC R85C NP_068758.3
XM_011525099.2 578 Missense Mutation CGC,TGC R85C XP_011523401.1
XM_011525100.2 578 Intron XP_011523402.1
Gene
NT5C3B
Gene Name
5'-nucleotidase, cytosolic IIIB
There are no transcripts associated with this gene.

Gene
P3H4
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
There are no transcripts associated with this gene.

View Full Product Details