Product Details

SNP ID
rs150709546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78358429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCTTTACCCCTTAAAGCGGGG[A/C]ATCGTACTGGTCCAGGAACTCCCGA
Phenotype
MIM: 604176
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928674 PubMed Links

Gene Details

Gene
LOC101928674
Gene Name
uncharacterized LOC101928674
There are no transcripts associated with this gene.

Gene
SOCS3
Gene Name
suppressor of cytokine signaling 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003955.4 1085 Missense Mutation GCC,TCC A223S NP_003946.3

View Full Product Details