Product Details

SNP ID
rs142649735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50269019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAAATTCCATGATAAAGTTGGAA[A/G]TCCATTCACCATTTGTAATACATAT
Phenotype
MIM: 614759 MIM: 156535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
There are no transcripts associated with this gene.

Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2808 UTR 3 NP_001191065.1
NM_001204137.1 2808 Intron NP_001191066.1
NM_001204138.1 2808 Intron NP_001191067.1
NM_001204139.1 2808 Intron NP_001191068.1
NM_001204140.1 2808 Intron NP_001191069.1
NM_001204141.1 2808 Intron NP_001191070.1
NM_001204142.1 2808 UTR 3 NP_001191071.1
NM_001204143.1 2808 UTR 3 NP_001191072.1
NM_001204151.2 2808 UTR 3 NP_001191080.1
NM_001323942.1 2808 UTR 3 NP_001310871.1
NM_001323947.1 2808 UTR 3 NP_001310876.1
NM_001323949.1 2808 UTR 3 NP_001310878.1
NM_001323950.1 2808 UTR 3 NP_001310879.1
NM_001323951.1 2808 Intron NP_001310880.1
NM_001323952.1 2808 UTR 3 NP_001310881.1
NM_001323953.1 2808 Intron NP_001310882.1
NM_001323954.1 2808 UTR 3 NP_001310883.1
NM_002384.2 2808 UTR 3 NP_002375.1
NM_015844.2 2808 UTR 3 NP_056669.2
NM_015845.3 2808 UTR 3 NP_056670.2
NM_015846.3 2808 UTR 3 NP_056671.2
NM_015847.3 2808 UTR 3 NP_056723.2
XM_005258271.2 2808 UTR 3 XP_005258328.1
XM_006722456.2 2808 UTR 3 XP_006722519.1
XM_011525991.1 2808 UTR 3 XP_011524293.1
XM_011525993.2 2808 UTR 3 XP_011524295.1
XM_011525994.2 2808 UTR 3 XP_011524296.1
XM_011525998.2 2808 Intron XP_011524300.1
XM_011525999.1 2808 UTR 3 XP_011524301.1
XM_011526001.1 2808 UTR 3 XP_011524303.1
XM_011526002.1 2808 UTR 3 XP_011524304.1
XM_011526003.2 2808 UTR 3 XP_011524305.1
XM_011526006.1 2808 UTR 3 XP_011524308.1
XM_011526007.1 2808 UTR 3 XP_011524309.1
XM_017025751.1 2808 UTR 3 XP_016881240.1
XM_017025752.1 2808 UTR 3 XP_016881241.1
XM_017025753.1 2808 UTR 3 XP_016881242.1
XM_017025754.1 2808 Intron XP_016881243.1
XM_017025755.1 2808 Intron XP_016881244.1
XM_017025756.1 2808 UTR 3 XP_016881245.1
XM_017025757.1 2808 UTR 3 XP_016881246.1
XM_017025758.1 2808 UTR 3 XP_016881247.1
XM_017025759.1 2808 UTR 3 XP_016881248.1
XM_017025760.1 2808 UTR 3 XP_016881249.1
XM_017025761.1 2808 Intron XP_016881250.1
XM_017025762.1 2808 UTR 3 XP_016881251.1
XM_017025763.1 2808 UTR 3 XP_016881252.1
XM_017025764.1 2808 UTR 3 XP_016881253.1
XM_017025765.1 2808 UTR 3 XP_016881254.1
XM_017025766.1 2808 UTR 3 XP_016881255.1
XM_017025767.1 2808 UTR 3 XP_016881256.1
XM_017025768.1 2808 Intron XP_016881257.1
XM_017025769.1 2808 UTR 3 XP_016881258.1
XM_017025770.1 2808 UTR 3 XP_016881259.1
XM_017025771.1 2808 UTR 3 XP_016881260.1
XM_017025772.1 2808 Intron XP_016881261.1
XM_017025773.1 2808 UTR 3 XP_016881262.1
XM_017025774.1 2808 UTR 3 XP_016881263.1
XM_017025775.1 2808 Intron XP_016881264.1
XM_017025776.1 2808 UTR 3 XP_016881265.1
XM_017025777.1 2808 Intron XP_016881266.1

View Full Product Details