Product Details

SNP ID
rs143072804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50269822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGAGTGCCTGCCCTGCAGACTT[C/G]AAGCTCCAGCATTAGATGCAAAGAC
Phenotype
MIM: 614759 MIM: 156535
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
There are no transcripts associated with this gene.

Gene
MBD1
Gene Name
methyl-CpG binding domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204136.1 2279 Intron NP_001191065.1
NM_001204137.1 2279 Intron NP_001191066.1
NM_001204138.1 2279 Intron NP_001191067.1
NM_001204139.1 2279 Intron NP_001191068.1
NM_001204140.1 2279 Intron NP_001191069.1
NM_001204141.1 2279 Intron NP_001191070.1
NM_001204142.1 2279 UTR 3 NP_001191071.1
NM_001204143.1 2279 Intron NP_001191072.1
NM_001204151.2 2279 UTR 3 NP_001191080.1
NM_001323942.1 2279 Intron NP_001310871.1
NM_001323947.1 2279 Intron NP_001310876.1
NM_001323949.1 2279 Intron NP_001310878.1
NM_001323950.1 2279 Intron NP_001310879.1
NM_001323951.1 2279 Intron NP_001310880.1
NM_001323952.1 2279 Intron NP_001310881.1
NM_001323953.1 2279 Intron NP_001310882.1
NM_001323954.1 2279 Intron NP_001310883.1
NM_002384.2 2279 Intron NP_002375.1
NM_015844.2 2279 Intron NP_056669.2
NM_015845.3 2279 Intron NP_056670.2
NM_015846.3 2279 Intron NP_056671.2
NM_015847.3 2279 Intron NP_056723.2
XM_005258271.2 2279 UTR 3 XP_005258328.1
XM_006722456.2 2279 Intron XP_006722519.1
XM_011525991.1 2279 Intron XP_011524293.1
XM_011525993.2 2279 Intron XP_011524295.1
XM_011525994.2 2279 Intron XP_011524296.1
XM_011525998.2 2279 Intron XP_011524300.1
XM_011525999.1 2279 Intron XP_011524301.1
XM_011526001.1 2279 Intron XP_011524303.1
XM_011526002.1 2279 UTR 3 XP_011524304.1
XM_011526003.2 2279 Intron XP_011524305.1
XM_011526006.1 2279 Intron XP_011524308.1
XM_011526007.1 2279 UTR 3 XP_011524309.1
XM_017025751.1 2279 Intron XP_016881240.1
XM_017025752.1 2279 Intron XP_016881241.1
XM_017025753.1 2279 Intron XP_016881242.1
XM_017025754.1 2279 Intron XP_016881243.1
XM_017025755.1 2279 Intron XP_016881244.1
XM_017025756.1 2279 Intron XP_016881245.1
XM_017025757.1 2279 UTR 3 XP_016881246.1
XM_017025758.1 2279 Intron XP_016881247.1
XM_017025759.1 2279 Intron XP_016881248.1
XM_017025760.1 2279 Intron XP_016881249.1
XM_017025761.1 2279 Intron XP_016881250.1
XM_017025762.1 2279 Intron XP_016881251.1
XM_017025763.1 2279 UTR 3 XP_016881252.1
XM_017025764.1 2279 Intron XP_016881253.1
XM_017025765.1 2279 Intron XP_016881254.1
XM_017025766.1 2279 Intron XP_016881255.1
XM_017025767.1 2279 Intron XP_016881256.1
XM_017025768.1 2279 Intron XP_016881257.1
XM_017025769.1 2279 Intron XP_016881258.1
XM_017025770.1 2279 UTR 3 XP_016881259.1
XM_017025771.1 2279 Intron XP_016881260.1
XM_017025772.1 2279 Intron XP_016881261.1
XM_017025773.1 2279 UTR 3 XP_016881262.1
XM_017025774.1 2279 Intron XP_016881263.1
XM_017025775.1 2279 Intron XP_016881264.1
XM_017025776.1 2279 UTR 3 XP_016881265.1
XM_017025777.1 2279 Intron XP_016881266.1

View Full Product Details