Product Details

SNP ID
rs139968960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10833115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGAAGTTGACCCGCTCCAAGTT[A/G]CCCTCTTGCAGGTTGCGGTCACGTG
Phenotype
MIM: 602378 MIM: 605395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf38 PubMed Links

Gene Details

Gene
C19orf38
Gene Name
chromosome 19 open reading frame 38
There are no transcripts associated with this gene.

Gene
DNM2
Gene Name
dynamin 2
There are no transcripts associated with this gene.

Gene
MIR6793
Gene Name
microRNA 6793
There are no transcripts associated with this gene.

Gene
TMED1
Gene Name
transmembrane p24 trafficking protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006858.3 680 Silent Mutation GGC,GGT G188G NP_006849.1
XM_006722631.3 680 Silent Mutation GGC,GGT G150G XP_006722694.1

View Full Product Details