Product Details
- SNP ID
-
rs140242272
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39705908 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGAAATCAATCATTCCCTCCAGTT[A/G]TGTCCCTGACCCACAGGCTTCATTT
- Phenotype
-
MIM: 607260
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS14
PubMed Links
Gene Details
- Gene
- LGALS14
- Gene Name
- galectin 14
View Full Product Details