Product Details

SNP ID
rs140427050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3227425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGGCTGTTTCTGGGAGCCCTGG[C/T]GTGGTTTAGTGTGCCTGTTCAGAGT
Phenotype
MIM: 612680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELF5 PubMed Links

Gene Details

Gene
CELF5
Gene Name
CUGBP, Elav-like family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172673.1 Intron NP_001166144.1
NM_021938.3 Intron NP_068757.2
XM_006722832.1 Intron XP_006722895.1
XM_006722833.1 Intron XP_006722896.1
XM_006722836.2 Intron XP_006722899.1
XM_006722837.3 Intron XP_006722900.1
XM_006722838.3 Intron XP_006722901.1
XM_011528173.1 Intron XP_011526475.1
XM_011528174.2 Intron XP_011526476.1
XM_017027111.1 Intron XP_016882600.1

View Full Product Details