Product Details

SNP ID
rs140704699
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3806122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGGAAGGCCAGCATTCGCAGGG[C/T]GTGCTGCGGGGCACACACAGCCTGT
Phenotype
MIM: 600038
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MATK PubMed Links

Gene Details

Gene
MATK
Gene Name
megakaryocyte-associated tyrosine kinase
There are no transcripts associated with this gene.

Gene
ZFR2
Gene Name
zinc finger RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145640.1 2661 Intron NP_001139112.1
NM_015174.1 2661 Missense Mutation ACC,GCC T883A NP_055989.1
XM_011527831.2 2661 Missense Mutation ACC,GCC T884A XP_011526133.1

View Full Product Details