Product Details

SNP ID
rs141556770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39605194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTCAGCAATGACCCACAGCTG[C/T]AGGTGGATTTCTACACTGACATGGA
Phenotype
MIM: 608717
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS13 PubMed Links

Gene Details

Gene
LGALS13
Gene Name
galectin 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013268.2 149 Nonsense Mutation CAG,TAG Q37* NP_037400.1
XM_011526874.2 149 Nonsense Mutation CAG,TAG Q30* XP_011525176.1
XM_017026715.1 149 Nonsense Mutation CAG,TAG Q68* XP_016882204.1

View Full Product Details