Product Details

SNP ID
rs143155018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35271110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTGATGGAACCAGAACACCCCG[A/T]GATGAGAGGAGAAGAGCCCAGCACA
Phenotype
MIM: 616582 MIM: 600390
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LSR PubMed Links

Gene Details

Gene
LSR
Gene Name
lipolysis stimulated lipoprotein receptor
There are no transcripts associated with this gene.

Gene
USF2
Gene Name
upstream transcription factor 2, c-fos interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321150.1 719 Silent Mutation CGA,CGT R101R NP_001308079.1
NM_003367.3 719 Silent Mutation CGA,CGT R232R NP_003358.1
NM_207291.2 719 Silent Mutation CGA,CGT R165R NP_997174.1
XM_005259197.3 719 Silent Mutation CGA,CGT R232R XP_005259254.1
XM_011527260.2 719 Silent Mutation CGA,CGT R232R XP_011525562.1
XM_011527261.2 719 Silent Mutation CGA,CGT R232R XP_011525563.1
XM_017027199.1 719 Silent Mutation CGA,CGT R165R XP_016882688.1

View Full Product Details