Product Details

SNP ID
rs144091748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11722547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATATCTTACATTTATGTGGTCC[A/G]TCTCCAGTGTGCCTTATCATGTGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 1050 Silent Mutation GAC,GAT D329D NP_001073962.1
NM_001297610.1 1050 Silent Mutation GAC,GAT D147D NP_001284539.1
NM_017507.1 1050 Silent Mutation GAC,GAT D285D NP_059977.1
XM_017026941.1 1050 Silent Mutation GAC,GAT D147D XP_016882430.1
XM_017026942.1 1050 Silent Mutation GAC,GAT D147D XP_016882431.1

View Full Product Details