Product Details

SNP ID
rs144274503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34486200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCCTCTCATTCCTGTAATCGC[C/T]GTCTGCCCAGCAGAACCCCGACTCT
Phenotype
MIM: 614790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WTIP PubMed Links

Gene Details

Gene
WTIP
Gene Name
Wilms tumor 1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080436.1 Intron NP_001073905.1
XM_006723014.3 Intron XP_006723077.1
XM_011526452.2 Intron XP_011524754.1
XM_011526453.2 Intron XP_011524755.1

View Full Product Details