Product Details

SNP ID
rs145694802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42868102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACGATGCTGGGATCCACTTACC[A/G]GAGACTTCGACTGTCATGGATTTGG
Phenotype
MIM: 176390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSG1 PubMed Links

Gene Details

Gene
PSG1
Gene Name
pregnancy specific beta-1-glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184825.1 1380 Silent Mutation TCC,TCT S414S NP_001171754.1
NM_001184826.1 1380 Silent Mutation TCC,TCT S414S NP_001171755.1
NM_001297773.1 1380 Silent Mutation TCC,TCT S414S NP_001284702.1
NM_006905.2 1380 Silent Mutation TCC,TCT S414S NP_008836.2
XM_005259065.3 1380 Silent Mutation TCC,TCT S321S XP_005259122.1
XM_011527125.1 1380 Silent Mutation TCC,TCT S321S XP_011525427.1
XM_017026994.1 1380 Silent Mutation TCC,TCT S321S XP_016882483.1

View Full Product Details