Product Details

SNP ID
rs145730490
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46648577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGATTTTCACGAAGACTTTGG[C/T]GGGGCCTGCGGGCGCCCCTGCACCT
Phenotype
MIM: 611112
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DACT3 PubMed Links

Gene Details

Gene
DACT3
Gene Name
dishevelled binding antagonist of beta catenin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301046.1 2645 Missense Mutation ACC,GCC T374A NP_001287975.1
NM_145056.2 2645 Missense Mutation ACC,GCC T599A NP_659493.2
XM_011526499.2 2645 Missense Mutation ACC,GCC T374A XP_011524801.1
XM_011526500.2 2645 Missense Mutation ACC,GCC T374A XP_011524802.1
XM_017026334.1 2645 Missense Mutation ACC,GCC T531A XP_016881823.1
Gene
GNG8
Gene Name
G protein subunit gamma 8
There are no transcripts associated with this gene.

View Full Product Details