Product Details

SNP ID
rs148737775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11722637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTACATGCATAGGGTTGTTCTCC[C/T]GTGTGAGTCCTTTCATGTAGTCGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF823 PubMed Links

Gene Details

Gene
ZNF823
Gene Name
zinc finger protein 823
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080493.3 960 Silent Mutation ACA,ACG T299T NP_001073962.1
NM_001297610.1 960 Silent Mutation ACA,ACG T117T NP_001284539.1
NM_017507.1 960 Silent Mutation ACA,ACG T255T NP_059977.1
XM_017026941.1 960 Silent Mutation ACA,ACG T117T XP_016882430.1
XM_017026942.1 960 Silent Mutation ACA,ACG T117T XP_016882431.1

View Full Product Details