Product Details

SNP ID
rs149108715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40777008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGGAGATCTGGCTGCTCGCCTG[G/T]GCTATTTCCCCAGTAGCATTGTCCG
Phenotype
MIM: 601340 MIM: 612945
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIA PubMed Links

Gene Details

Gene
MIA
Gene Name
melanoma inhibitory activity
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202553.1 467 Missense Mutation GGC,TGC G101C NP_001189482.1
NM_006533.3 467 Missense Mutation GGC,TGC G101C NP_006524.1
Gene
MIA-RAB4B
Gene Name
MIA-RAB4B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
RAB4B
Gene Name
RAB4B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
RAB4B-EGLN2
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details