Product Details
- SNP ID
-
rs150028589
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49665959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCTACGCTGGGCCGGTTATCGA[C/G]CCGGCCCAGTGCGCAGGCGCGGGAA
- Phenotype
-
MIM: 610837
MIM: 603734
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BCL2L12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2060263] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCL2L12
- Gene Name
- BCL2 like 12
- Gene
- IRF3
- Gene Name
- interferon regulatory factor 3
- Gene
- SCAF1
- Gene Name
- SR-related CTD associated factor 1
There are no transcripts associated with this gene.
View Full Product Details