Product Details

SNP ID
rs112272202
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:157676653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAATTTGCACATTTGTTATATC[C/T]GAGATGTACAGTTAGGACTCACCCC
Phenotype
MIM: 606510
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCRL3 PubMed Links

Gene Details

Gene
FCRL3
Gene Name
Fc receptor like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320333.1 2595 UTR 3 NP_001307262.1
NM_052939.3 2595 Intron NP_443171.2
XM_006711145.1 2595 UTR 3 XP_006711208.1

View Full Product Details