Product Details

SNP ID
rs112462014
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:71064944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAATAAACACAGCTGTATTGTT[T/C]TGAAAAGCAATGAAAGGCATGCACC
Phenotype
MIM: 604347
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MIR186 PubMed Links

Gene Details

Gene
MIR186
Gene Name
microRNA 186
There are no transcripts associated with this gene.

Gene
ZRANB2
Gene Name
zinc finger RANBP2-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005455.4 1417 UTR 3 NP_005446.2
NM_203350.2 1417 UTR 3 NP_976225.1
Gene
ZRANB2-AS1
Gene Name
ZRANB2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details