Product Details

SNP ID
rs112655761
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:157677821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAACATGTAGATACATTAATAT[G/A]ATAGAAATAGGAGAGCATGGGAATA
Phenotype
MIM: 606510
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
FCRL3 PubMed Links

Gene Details

Gene
FCRL3
Gene Name
Fc receptor like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320333.1 3316 Intron NP_001307262.1
NM_052939.3 3316 Intron NP_443171.2
XM_006711145.1 3316 UTR 3 XP_006711208.1

View Full Product Details