Product Details

SNP ID
rs113193158
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:243256170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCAGCTCTGGGCCTAGAGTGC[G/A]TGCATGGCGAAGTCCCCGGAGAACT
Phenotype
MIM: 613023 MIM: 613524
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CEP170 PubMed Links

Gene Details

Gene
CEP170
Gene Name
centrosomal protein 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042404.1 121 Intron NP_001035863.1
NM_001042405.1 121 Intron NP_001035864.1
NM_014812.2 121 Intron NP_055627.2
XM_006711843.3 121 Intron XP_006711906.1
XM_011544334.2 121 Intron XP_011542636.1
XM_011544335.2 121 Intron XP_011542637.1
XM_011544336.2 121 Intron XP_011542638.1
XM_011544337.2 121 Intron XP_011542639.1
XM_011544338.2 121 Intron XP_011542640.1
XM_011544339.2 121 Intron XP_011542641.1
XM_011544340.2 121 Intron XP_011542642.1
XM_011544341.2 121 Intron XP_011542643.1
XM_011544342.2 121 Intron XP_011542644.1
XM_011544343.2 121 Intron XP_011542645.1
XM_011544344.2 121 Intron XP_011542646.1
XM_017002932.1 121 Intron XP_016858421.1
XM_017002933.1 121 Intron XP_016858422.1
XM_017002934.1 121 Intron XP_016858423.1
XM_017002935.1 121 Intron XP_016858424.1
XM_017002936.1 121 Intron XP_016858425.1
XM_017002937.1 121 Intron XP_016858426.1
XM_017002938.1 121 Intron XP_016858427.1
XM_017002939.1 121 Intron XP_016858428.1
XM_017002940.1 121 Intron XP_016858429.1
XM_017002941.1 121 Intron XP_016858430.1
XM_017002942.1 121 Intron XP_016858431.1
XM_017002943.1 121 Intron XP_016858432.1
XM_017002944.1 121 Intron XP_016858433.1
XM_017002945.1 121 Intron XP_016858434.1
XM_017002946.1 121 Intron XP_016858435.1
XM_017002947.1 121 Intron XP_016858436.1
XM_017002948.1 121 Intron XP_016858437.1
XM_017002949.1 121 Intron XP_016858438.1
XM_017002950.1 121 Intron XP_016858439.1
XM_017002951.1 121 Intron XP_016858440.1
Gene
SDCCAG8
Gene Name
serologically defined colon cancer antigen 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006642.3 121 UTR 5 NP_006633.1
XM_005273013.4 121 UTR 5 XP_005273070.1
XM_005273018.1 121 UTR 5 XP_005273075.1
XM_005273021.4 121 Intron XP_005273078.1
XM_005273022.3 121 UTR 5 XP_005273079.1
XM_005273023.4 121 UTR 5 XP_005273080.1
XM_006711727.3 121 Intron XP_006711790.1
XM_006711728.3 121 Intron XP_006711791.1
XM_011544021.2 121 Intron XP_011542323.1
XM_011544022.2 121 UTR 5 XP_011542324.1
XM_011544023.2 121 Intron XP_011542325.1
XM_011544024.2 121 Intron XP_011542326.1
XM_011544026.2 121 Intron XP_011542328.1
XM_011544028.2 121 Intron XP_011542330.1
XM_011544029.2 121 Intron XP_011542331.1
XM_011544030.2 121 Intron XP_011542332.1
XM_017000103.1 121 Intron XP_016855592.1
XM_017000104.1 121 UTR 5 XP_016855593.1
XM_017000105.1 121 UTR 5 XP_016855594.1
XM_017000106.1 121 UTR 5 XP_016855595.1
XM_017000107.1 121 Intron XP_016855596.1
XM_017000108.1 121 UTR 5 XP_016855597.1
XM_017000109.1 121 Intron XP_016855598.1

View Full Product Details