Product Details

SNP ID
rs113394969
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:11787280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCACATGGGGGAATGGGTGGAA[-/GAG]GAGGCCGGGATGTGTCCTGCCAGC
Phenotype
MIM: 607093
Polymorphism
-/GAG, Insertion/deletion
Allele Nomenclature
Literature Links
C1orf167 PubMed Links

Gene Details

Gene
C1orf167
Gene Name
chromosome 1 open reading frame 167
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010881.1 5599 Intron NP_001010881.1
XM_011541269.2 5599 Intron XP_011539571.2
XM_011541271.2 5599 Intron XP_011539573.2
XM_011541272.2 5599 Intron XP_011539574.1
XM_011541274.2 5599 Intron XP_011539576.2
XM_011541275.2 5599 Intron XP_011539577.2
XM_011541276.2 5599 Intron XP_011539578.1
XM_011541277.2 5599 Intron XP_011539579.1
XM_011541278.2 5599 Intron XP_011539580.1
XM_011541280.2 5599 Intron XP_011539582.2
XM_011541281.2 5599 Intron XP_011539583.2
XM_017001062.1 5599 Intron XP_016856551.1
XM_017001063.1 5599 Intron XP_016856552.1
XM_017001064.1 5599 Intron XP_016856553.1
Gene
LOC102724659
Gene Name
uncharacterized LOC102724659
There are no transcripts associated with this gene.

Gene
MTHFR
Gene Name
methylenetetrahydrofolate reductase (NAD(P)H)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005957.4 5599 UTR 3 NP_005948.3
XM_005263458.3 5599 Intron XP_005263515.1
XM_005263460.4 5599 Intron XP_005263517.1
XM_005263462.4 5599 Intron XP_005263519.1
XM_005263463.3 5599 Intron XP_005263520.1
XM_011541495.2 5599 Intron XP_011539797.1
XM_011541496.2 5599 Intron XP_011539798.1
XM_017001328.1 5599 Intron XP_016856817.1

View Full Product Details