Product Details

SNP ID
rs113399506
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:227921078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCACAGCAGGGTTGGCCAGGCC[C/T]GGGGACTCGTCCCTTGCAGGTGTAG
Phenotype
MIM: 602863
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT9A PubMed Links

Gene Details

Gene
WNT9A
Gene Name
Wnt family member 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003395.2 2109 Intron NP_003386.1
XM_011544271.2 2109 UTR 3 XP_011542573.1

View Full Product Details