Product Details

SNP ID
rs114339113
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:200551570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGTCAGTGCACATAATTCCAA[T/C]AGCAAATATTTTTTCACCATTCATT
Phenotype
MIM: 611279
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
KIF14 PubMed Links

Gene Details

Gene
KIF14
Gene Name
kinesin family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305792.1 6516 UTR 3 NP_001292721.1
NM_014875.2 6516 UTR 3 NP_055690.1
XM_011510231.2 6516 UTR 3 XP_011508533.1
XM_011510232.2 6516 UTR 3 XP_011508534.1
XM_011510233.2 6516 UTR 3 XP_011508535.1
XM_011510235.2 6516 UTR 3 XP_011508537.1
XM_011510236.1 6516 UTR 3 XP_011508538.1
XM_017003005.1 6516 UTR 3 XP_016858494.1
XM_017003006.1 6516 UTR 3 XP_016858495.1
XM_017003007.1 6516 UTR 3 XP_016858496.1

View Full Product Details