Product Details

SNP ID
rs114432947
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:177929741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTAAGTGCCCGGTGGAGGCAT[C/T]GGGCTAGCACAAACCTCTTGAGGGT
Phenotype
MIM: 612855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC16B PubMed Links

Gene Details

Gene
SEC16B
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033127.2 3390 UTR 3 NP_149118.2

View Full Product Details