Product Details

SNP ID
rs114500445
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:68474959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCACATTTTTGCTAAGATCTC[T/C]TCTAGTGCTATAATCCTAGGTGATT
Phenotype
MIM: 612002
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DEPDC1 PubMed Links

Gene Details

Gene
DEPDC1
Gene Name
DEP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114120.2 4631 UTR 3 NP_001107592.1
NM_017779.5 4631 UTR 3 NP_060249.2

View Full Product Details