Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319179.1 | 1880 | Missense Mutation | NP_001306108.1 | ||
NM_001319180.1 | 1880 | Missense Mutation | NP_001306109.1 | ||
NM_001319181.1 | 1880 | Intron | NP_001306110.1 | ||
NM_018284.2 | 1880 | Missense Mutation | NP_060754.2 | ||
XM_005270743.2 | 1880 | Missense Mutation | XP_005270800.1 | ||
XM_006710574.2 | 1880 | Missense Mutation | XP_006710637.1 | ||
XM_011541233.1 | 1880 | Missense Mutation | XP_011539535.1 | ||
XM_011541234.1 | 1880 | Missense Mutation | XP_011539536.1 | ||
XM_017001013.1 | 1880 | Missense Mutation | XP_016856502.1 | ||
XM_017001014.1 | 1880 | Missense Mutation | XP_016856503.1 |