Product Details

SNP ID
rs115547854
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:166603919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGTTGTCTCAAAGTCATCATA[C/T]GGAAAACAGAATAGAAGCACAGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMO9P PubMed Links

Gene Details

Gene
FMO9P
Gene Name
flavin containing monooxygenase 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details