Product Details

SNP ID
rs116181846
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:200874064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCCTCCTCGCTCTCCAGGGAC[G/T]ACAGTTCCGTGTTCCGTTGCCGGGC
Phenotype
MIM: 602174
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 1110 Missense Mutation GAC,TAC D343Y NP_005289.2

View Full Product Details