Product Details

SNP ID
rs116223306
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:109625658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGCTGACCTTCCTTCCCTCCCC[C/T]CAGGAGCTGTTCACCCGCTCACTGG
Phenotype
MIM: 102771 MIM: 139340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMPD2 PubMed Links

Gene Details

Gene
AMPD2
Gene Name
adenosine monophosphate deaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257360.1 232 Intron NP_001244289.1
NM_001257361.1 232 Intron NP_001244290.1
NM_001308170.1 232 Intron NP_001295099.1
NM_004037.7 232 Intron NP_004028.3
NM_139156.3 232 Intron NP_631895.1
NM_203404.1 232 Silent Mutation CCC,CCT P8P NP_981949.1
Gene
GNAT2
Gene Name
G protein subunit alpha transducin 2
There are no transcripts associated with this gene.

View Full Product Details