Product Details

SNP ID
rs116448589
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:59308150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCCTCTTAATGCTGCTGCTTCT[C/T]TTTTTATTTTAGAAAACCATGTCAT
Phenotype
MIM: 611370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGGY PubMed Links

Gene Details

Gene
FGGY
Gene Name
FGGY carbohydrate kinase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113411.1 Intron NP_001106882.1
NM_001244714.1 Intron NP_001231643.1
NM_001278224.1 Intron NP_001265153.1
NM_018291.3 Intron NP_060761.3
XM_011541730.1 Intron XP_011540032.1
XM_011541731.1 Intron XP_011540033.1
XM_011541732.1 Intron XP_011540034.1
XM_011541733.1 Intron XP_011540035.1
XM_011541735.1 Intron XP_011540037.1
XM_011541736.2 Intron XP_011540038.1
XM_017001643.1 Intron XP_016857132.1
XM_017001644.1 Intron XP_016857133.1
XM_017001645.1 Intron XP_016857134.1
XM_017001646.1 Intron XP_016857135.1
XM_017001647.1 Intron XP_016857136.1
XM_017001648.1 Intron XP_016857137.1
XM_017001649.1 Intron XP_016857138.1
XM_017001650.1 Intron XP_016857139.1
XM_017001651.1 Intron XP_016857140.1
XM_017001652.1 Intron XP_016857141.1
XM_017001653.1 Intron XP_016857142.1
XM_017001654.1 Intron XP_016857143.1
XM_017001655.1 Intron XP_016857144.1
XM_017001656.1 Intron XP_016857145.1
XM_017001657.1 Intron XP_016857146.1
XM_017001658.1 Intron XP_016857147.1
XM_017001659.1 Intron XP_016857148.1
XM_017001660.1 Intron XP_016857149.1
XM_017001661.1 Intron XP_016857150.1
XM_017001662.1 Intron XP_016857151.1
XM_017001663.1 Intron XP_016857152.1
XM_017001664.1 Intron XP_016857153.1
XM_017001665.1 Intron XP_016857154.1
XM_017001666.1 Intron XP_016857155.1
XM_017001667.1 Intron XP_016857156.1
XM_017001668.1 Intron XP_016857157.1
XM_017001669.1 Intron XP_016857158.1
XM_017001670.1 Intron XP_016857159.1
XM_017001671.1 Intron XP_016857160.1
XM_017001672.1 Intron XP_016857161.1
XM_017001673.1 Intron XP_016857162.1
XM_017001674.1 Intron XP_016857163.1
XM_017001675.1 Intron XP_016857164.1
XM_017001676.1 Intron XP_016857165.1
XM_017001677.1 Intron XP_016857166.1
XM_017001678.1 Intron XP_016857167.1
XM_017001679.1 Intron XP_016857168.1

View Full Product Details