Product Details

SNP ID
rs116531456
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:145742338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTTGTCAGAAAAATTAAAAAA[G/A]ATCAATAGATAACCGTATTGTAGTT
Phenotype
MIM: 604463
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CD160 PubMed Links

Gene Details

Gene
CD160
Gene Name
CD160 molecule
There are no transcripts associated with this gene.

Gene
RNF115
Gene Name
ring finger protein 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014455.3 Intron NP_055270.1
XM_005272952.4 Intron XP_005273009.1

View Full Product Details