Product Details

SNP ID
rs116714084
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:91643071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCTTAAGGTGATTAGGAAGAAG[C/A]TTGTTTGTAAAACTCTGGACAAGAT
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
HSP90B3P PubMed Links

Gene Details

Gene
HSP90B3P
Gene Name
heat shock protein 90 beta family member 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details