Product Details

SNP ID
rs117133279
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:221701462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAATGAAAATAAATTTTTACACA[A/G]TCAACAGAAACACACCAAGATTTTT
Phenotype
MIM: 608867
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP10 PubMed Links

Gene Details

Gene
DUSP10
Gene Name
dual specificity phosphatase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007207.5 2638 UTR 3 NP_009138.1
XM_017000147.1 2638 Intron XP_016855636.1

View Full Product Details