Product Details

SNP ID
rs117599057
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:93152211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGCAAACTCTAGGCCTTAGTG[T/G]GAGGTGAACACGGTAACTGCTACAT
Phenotype
MIM: 616876
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 3645 UTR 3 NP_001161302.1
NM_016040.4 3645 UTR 3 NP_057124.3

View Full Product Details