Product Details

SNP ID
rs138366687
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:159782262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAGATGCCATTGCTGAAATCC[A/G]ACGACTACGACCCGGCTCCATCGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP23 PubMed Links

Gene Details

Gene
DUSP23
Gene Name
dual specificity phosphatase 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319658.1 427 Missense Mutation CAA,CGA Q126R NP_001306587.1
NM_001319659.1 427 Missense Mutation CAA,CGA Q126R NP_001306588.1
NM_017823.4 427 Missense Mutation CAA,CGA Q126R NP_060293.2

View Full Product Details