Product Details

SNP ID
rs140345484
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:193180842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACATATGGAATATGTGGACAGT[A/G]TGTTGCAACCCAGTTCATGCCCATT
Phenotype
MIM: 603018 MIM: 607393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GALT2 PubMed Links

Gene Details

Gene
B3GALT2
Gene Name
beta-1,3-galactosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003783.3 1436 Missense Mutation CAC,TAC H241Y NP_003774.1
Gene
CDC73
Gene Name
cell division cycle 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024529.4 1436 Intron NP_078805.3
XM_006711537.3 1436 Intron XP_006711600.1

View Full Product Details