Product Details

SNP ID
rs141591932
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:54174458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGATTAACCTCATGGGGCAGAT[G/A]AGAAGTAGAGGTTCTGAGCAGTAAA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CYB5RL PubMed Links

Gene Details

Gene
CYB5RL
Gene Name
cytochrome b5 reductase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031672.2 1429 UTR 3 NP_001026842.2

View Full Product Details