Product Details

SNP ID
rs143842871
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:95093941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATATGACCCTGTGCAATGTGAC[C/T]TTTCTGTTTTTCTCATTAAGAGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM56 PubMed Links

Gene Details

Gene
TMEM56
Gene Name
transmembrane protein 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199679.1 Intron NP_001186608.1
NM_152487.2 Intron NP_689700.1

View Full Product Details