Product Details
- SNP ID
-
rs143902744
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:89007747 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTAGATCTTTAGCTTATGCGACA[C/T]ATATCTCTTGGTTTTTTTTTTCAGG
- Phenotype
-
MIM: 600413
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GBP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs548474277] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GBP3
- Gene Name
- guanylate binding protein 3
View Full Product Details