Product Details

SNP ID
rs144004857
Assay Type
Functionally Tested
NCBI dbSNP Submissions
10
Location
Chr.1:206798930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGCGTTTCCGCACAGATCTGC[A/G]TGTTCCTTACCACTCACACATGTGC
Phenotype
MIM: 605687
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IL19 PubMed Links

Gene Details

Gene
IL19
Gene Name
interleukin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013371.3 61 Intron NP_037503.2
NM_153758.2 61 Silent Mutation GCA,GCG A12A NP_715639.1
XM_011509450.2 61 Intron XP_011507752.1

View Full Product Details