Product Details
- SNP ID
-
rs144204544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:160995928 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTATCCCAGATTATGCTGAGTTC[A/C]TGTCTCAGGGGAAACACCAAAATCC
- Phenotype
-
MIM: 605721
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
F11R
PubMed Links
Gene Details
- Gene
- F11R
- Gene Name
- F11 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016946.4 |
4113 |
UTR 3 |
|
|
NP_058642.1 |
View Full Product Details