Product Details

SNP ID
rs144208698
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:44806028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTCGCAGTAGAGCCCGTGGACC[A/G]TGGCCACCGCGAAGAGCGAGGTGTT
Phenotype
MIM: 602913 MIM: 611713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 784 Intron NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 784 Missense Mutation ACG,ATG T214M NP_001013654.1
XM_006710614.3 784 Missense Mutation ACG,ATG T272M XP_006710677.1
XM_011541375.2 784 Missense Mutation ACG,ATG T214M XP_011539677.1
XM_011541376.2 784 Missense Mutation ACG,ATG T214M XP_011539678.1
XM_017001187.1 784 Missense Mutation ACG,ATG T255M XP_016856676.1

View Full Product Details