Product Details

SNP ID
rs145048286
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:153418208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGATGAAGGAGAACTTCCCCAA[C/T]TTCCTCAGTGCCTGTGTGAGTTGGG
Phenotype
MIM: 123885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
S100A7A PubMed Links

Gene Details

Gene
S100A7A
Gene Name
S100 calcium binding protein A7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176823.3 Intron NP_789793.1
Gene
S100A8
Gene Name
S100 calcium binding protein A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319196.1 Intron NP_001306125.1
NM_001319197.1 Intron NP_001306126.1
NM_001319198.1 Intron NP_001306127.1
NM_001319201.1 Intron NP_001306130.1
NM_002964.4 Intron NP_002955.2

View Full Product Details